All Photos(1)

AMBH581767DB

Ambeed, Inc.

4-Phenylsemicarbazide

CAS Number:
Molecular Weight:
151.17
MDL number:

mp

122-127�t N/A mmHg

Iupac Name

N-phenylhydrazinecarboxamide

Inchi Code

1S/C7H9N3O/c8-10-7(11)9-6-4-2-1-3-5-6/h1-5H,8H2,(H2,9,10,11)

InChI key

MOCKWYUCPREFCZ-UHFFFAOYSA-N

storage temp.

Keep in dark place,Inert atmosphere,Room temperature

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/537-47-3.html

Melting Point

122-127�t N/A mmHg

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

H315-H319-H335

MSDS

https://www.ambeed.com/products/537-47-3.html

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service