Sign In to View Organizational & Contract Pricing
Select a Size
1 G
$28.75
5 G
$97.75
10 G
$178.25
25 G
$327.75
Vendor SKU: A429277-1G
$28.75
Ships in 7 days - from Aldrich Partner.
About This Item
Skip To
Iupac Name
N-(tert-butyl)benzenesulfonamide
Inchi Code
1S/C10H15NO2S/c1-10(2,3)11-14(12,13)9-7-5-4-6-8-9/h4-8,11H,1-3H3
InChI key
FFUBXANSXRGVKW-UHFFFAOYSA-N
storage temp.
Sealed in dry, room temperature
Purity
96%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
solid
Product Link
https://www.ambeed.com/products/2512-24-5.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.ambeed.com/products/2512-24-5.html
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service