Skip to Content
MilliporeSigma

AMBH6FD7F9E2

Ambeed, Inc.

4-Bromo-1H-pyrrole-2-carbonitrile

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
171
MDL number:
Marketplace Support
Need help or have questions about this product?
Contact Seller
Marketplace Support
Need help or have questions about this product?
Contact Seller

Iupac Name

4-bromo-1H-pyrrole-2-carbonitrile

Inchi Code

1S/C5H3BrN2/c6-4-1-5(2-7)8-3-4/h1,3,8H

InChI key

MDGARDUARHMMAR-UHFFFAOYSA-N

storage temp.

Sealed in dry, 2-8C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/1221435-18-2.html

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.ambeed.com/products/1221435-18-2.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library