MilliporeSigma
All Photos(1)

AMBH6FD84091

Ambeed, Inc.

DL-Histidine monohydrochloride monohydrate

CAS Number:
Molecular Weight:
209.63
MDL number:

bp

458.9�t 760 mmHg

mp

253�t N/A mmHg

Iupac Name

histidine hydrochloride hydrate

Inchi Code

1S/C6H9N3O2.ClH.H2O/c7-5(6(10)11)1-4-2-8-3-9-4;;/h2-3,5H,1,7H2,(H,8,9)(H,10,11);1H;1H2

InChI key

CMXXUDSWGMGYLZ-UHFFFAOYSA-N

storage temp.

Inert atmosphere,Room Temperature

Purity

99%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/123333-71-1.html

Flash Point

231.3

Boiling point

458.9�t 760 mmHg

Melting Point

253�t N/A mmHg

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

H302-H315-H319-H335

Precautionary statements

MSDS

https://www.ambeed.com/products/123333-71-1.html

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service