MilliporeSigma
All Photos(1)

AMBH93E4C1EE

Ambeed, Inc.

2-(Methylsulfonyl) ethanol

CAS Number:
Molecular Weight:
124.16
MDL number:

bp

344.5�t 760 mmHg

mp

31-33�t N/A mmHg

Iupac Name

2-(methylsulfonyl)ethanol

Inchi Code

1S/C3H8O3S/c1-7(5,6)3-2-4/h4H,2-3H2,1H3

InChI key

KFTYFTKODBWKOU-UHFFFAOYSA-N

storage temp.

Sealed in dry,Room Temperature

Purity

97%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Colorless to Yellow Liquid or Semi-Solid or solid

Product Link

https://www.ambeed.com/products/15205-66-0.html

Flash Point

162.2

Boiling point

344.5�t 760 mmHg

Melting Point

31-33�t N/A mmHg

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

H302-H315-H319-H335

MSDS

https://www.ambeed.com/products/15205-66-0.html

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service