MilliporeSigma
All Photos(1)

AMBH93EDD0CA

Ambeed, Inc.

Dichloro bis(triethylphosphine) palladium(II)

CAS Number:
Molecular Weight:
415.66
MDL number:

Iupac Name

bis(triethyl-lambda5-phosphanyl)palladium(IV) chloride

Inchi Code

1S/2C6H15P.2ClH.Pd/c2*1-4-7(5-2)6-3;;;/h2*4-6H2,1-3H3;2*1H;

InChI key

YQLNDFXCSZLJNP-UHFFFAOYSA-N

storage temp.

Inert atmosphere,2-8

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/28425-04-9.html

Synonym

Bis(triethylphosphoranyl)palladium(IV) chloride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.ambeed.com/products/28425-04-9.html

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service