All Photos(1)

AMBH95E07FC5

Ambeed, Inc.

(2R)-3-(((2-Aminoethoxy)(hydroxy)phosphoryl)oxy)propane-1,2-diyl bis(decanoate)

CAS Number:
Molecular Weight:
523.65
MDL number:

Iupac Name

(2R)-3-(((2-aminoethoxy)(hydroxy)phosphoryl)oxy)propane-1,2-diyl bis(decanoate)

Inchi Code

1S/C25H50NO8P/c1-3-5-7-9-11-13-15-17-24(27)31-21-23(22-33-35(29,30)32-20-19-26)34-25(28)18-16-14-12-10-8-6-4-2/h23H,3-22,26H2,1-2H3,(H,29,30)/t23-/m1/s1

InChI key

KKOSJVWUOHEQKA-HSZRJFAPSA-N

storage temp.

Keep in dark place,Inert atmosphere,Store in freezer, under -20

Purity

95%

Country of Origin

CN

Shipping Temperature

Cold-chain transportation

Physical Form

solid

Product Link

https://www.ambeed.com/products/253685-27-7.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

H302-H315-H319-H335

Precautionary statements

MSDS

https://www.ambeed.com/products/253685-27-7.html

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service