Skip to Content
MilliporeSigma

AMBH9614F508

Ambeed, Inc.

2-Morpholino-5-(trifluoromethyl)aniline

Synonym(s):

2-(4-morpholinyl)-5-(trifluoromethyl)phenylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

250 MG

$28.75

1 G

$47.15

5 G

$113.85

25 G

$399.05

Vendor SKU: A307730-250MG

$28.75


Ships in 5 days - from Aldrich Partner.


About This Item

Linear Formula:
C11H13O1N2F3
CAS Number:
Molecular Weight:
246.23
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C11H13F3N2O/c12-11(13,14)8-1-2-10(9(15)7-8)16-3-5-17-6-4-16/h1-2,7H,3-6,15H2

Inchi Key

CNVOJNRNRNAOOP-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

solid

signalword

Danger

pictograms

Skull and crossbones

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service