All Photos(1)

AMBH97B9F79E

Ambeed, Inc.

3-Chloropropionaldehydediethylacetal

CAS Number:
Molecular Weight:
166.65
MDL number:

Iupac Name

3-chloro-1,1-diethoxypropane

Inchi Code

1S/C7H15ClO2/c1-3-9-7(5-6-8)10-4-2/h7H,3-6H2,1-2H3

InChI key

NXHONHDWVLPPCS-UHFFFAOYSA-N

storage temp.

Inert atmosphere,2-8

Purity

95%23

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.ambeed.com/products/35573-93-4.html

Pictograms

Flame

Signal Word

Danger

Hazard Statements

H226

Precautionary statements

MSDS

https://www.ambeed.com/products/35573-93-4.html

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service