MilliporeSigma
All Photos(1)

AMBH97BA09B4

Ambeed, Inc.

2-Amino-6-methylpteridin-4(3H)-one

CAS Number:
Molecular Weight:
177.17
MDL number:

Iupac Name

2-amino-6-methyl-4-pteridinol

Inchi Code

1S/C7H7N5O/c1-3-2-9-5-4(10-3)6(13)12-7(8)11-5/h2H,1H3,(H3,8,9,11,12,13)

InChI key

UDOGNMDURIJYQC-UHFFFAOYSA-N

storage temp.

Keep in dark place,Sealed in dry,2-8

Purity

95%

Country of Origin

US

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/708-75-8.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.ambeed.com/products/708-75-8.html

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service