MilliporeSigma
All Photos(1)

AMBH97BA0EB4

Ambeed, Inc.

(2S,4R)-N-((S)-2-(tert-Butyl)-17-((S)-4-(4-chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)-4,16-dioxo-6,9,12-trioxa-3,15-diazaheptadecan-1-oyl)-4-hydroxy-1-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide

CAS Number:
Molecular Weight:
1002.66
MDL number:

storage temp.

Sealed in dry,Store in freezer, under -20

Purity

98%

Country of Origin

CN

Shipping Temperature

Shipped in cold pack

Physical Form

solid

Product Link

https://www.ambeed.com/products/1797406-69-9.html

Synonym

(2S,4R)-N-((S)-2-(tert-Butyl)-17-((S)-4-(4-chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)-4,16-dioxo-6,9,12-trioxa-3,15-diazaheptadecan-1-oyl)-4-hydroxy-1-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

H302-H315-H319-H335

Precautionary statements

MSDS

https://www.ambeed.com/products/1797406-69-9.html

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service