Skip to Content
MilliporeSigma

AMBH97BA11D9

Ambeed, Inc.

3-Hydroxy-N-(2-methoxyphenyl)-2-naphthamide

Synonym(s):

3-hydroxy-N-(2-methoxyphenyl)-2-naphthamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Product AMBH97BA11D9 is currently not available for purchase. Contact Technical Service

About This Item

Linear Formula:
C18H15O3N1
CAS Number:
Molecular Weight:
293.32
MDL number:

Skip To

Inchi Code

1S/C18H15NO3/c1-22-17-9-5-4-8-15(17)19-18(21)14-10-12-6-2-3-7-13(12)11-16(14)20/h2-11,20H,1H3,(H,19,21)

Inchi Key

AQYMRQUYPFCXDM-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

solid


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library