Skip to Content
MilliporeSigma

AMBH97F03EF6

Ambeed, Inc.

5-Iodo-1-pentyne

Synonym(s):

5-iodo-1-pentyne

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
Molecular Weight:
194.02
MDL number:

Inchi Code

1S/C5H7I/c1-2-3-4-5-6/h1H,3-5H2

Inchi Key

SEHFYZRHGUPLSY-UHFFFAOYSA-N

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Purity

97%

Country of Origin

CN

Shipping Temperature

Cold-chain transportation

Physical Form

liquid


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library