Skip to Content
MilliporeSigma

AMBH97F0454C

Ambeed, Inc.

Bromophenol blue sodium

Synonym(s):

sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate

Sign In to View Organizational & Contract Pricing.

Select a Size

Product AMBH97F0454C is currently not available for purchase. Contact Technical Service

About This Item

Linear Formula:
C19H9O5Br4S1Na1
CAS Number:
Molecular Weight:
691.95
MDL number:

Skip To

Inchi Code

1S/C19H10Br4O5S.Na/c20-12-5-9(6-13(21)17(12)24)19(10-7-14(22)18(25)15(23)8-10)11-3-1-2-4-16(11)29(26,27)28-19;/h1-8,24-25H;/q;+1/p-1

Inchi Key

QDVHDLHGPSKICA-UHFFFAOYSA-M

Shipping Temperature

Normal

Purity

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

solid

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library