Skip to Content
MilliporeSigma

AMBH97F045A4

Ambeed, Inc.

Silver sulfide

Synonym(s):

disilver(1+) sulfide

Sign In to View Organizational & Contract Pricing.

Select a Size

1 G

$19.55

5 G

$40.25

25 G

$72.45

Vendor SKU: A1455827-1G

$19.55


Ships in 5 days - from Aldrich Partner.


Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/2Ag.S/q2*+1;-2

Inchi Key

XUARKZBEFFVFRG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

99.9%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

solid

Melting Point

845C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service