Skip to Content
MilliporeSigma

AMBH97F045C4

Ambeed, Inc.

Amaranth

Synonym(s):

trisodium 3-hydroxy-4-[(E)-(4-sulfonato-1-naphthyl)diazenyl]-2,7-naphthalenedisulfonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Product AMBH97F045C4 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
604.48
MDL number:

Skip To

Inchi Code

1S/C20H14N2O10S3.3Na/c23-20-18(35(30,31)32)10-11-9-12(33(24,25)26)5-6-13(11)19(20)22-21-16-7-8-17(34(27,28)29)15-4-2-1-3-14(15)16;;;/h1-10,23H,(H,24,25,26)(H,27,28,29)(H,30,31,32);;;/q;3*+1/p-3/b22-21+;;;

Inchi Key

WLDHEUZGFKACJH-ZRUFZDNISA-K

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Purity

85%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

MSDS

https://www.ambeed.com/products/915-67-3.html

hstatements

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library