MilliporeSigma
All Photos(1)

AMBH97F05C15

Ambeed, Inc.

1-Acetylindoline-2,3-dione

CAS Number:
Molecular Weight:
189.17
MDL number:

bp

345.4�t 760 mmHg

mp

141-143�t N/A mmHg

Iupac Name

1-acetyl-1H-indole-2,3-dione

Inchi Code

1S/C10H7NO3/c1-6(12)11-8-5-3-2-4-7(8)9(13)10(11)14/h2-5H,1H3

InChI key

LPGDEHBASRKTDG-UHFFFAOYSA-N

storage temp.

Sealed in dry,2-8

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/574-17-4.html

Flash Point

165.8

Boiling point

345.4�t 760 mmHg

Melting Point

141-143�t N/A mmHg

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.ambeed.com/products/574-17-4.html

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service