Skip to Content
MilliporeSigma

AMBH97F06F47

Ambeed, Inc.

GDP-D-mannose disodium

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Product AMBH97F06F47 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
649.31
MDL number:

Skip To

Marketplace Support
Need help or have questions about this product?
Contact Seller

Iupac Name

sodium 9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(((((hydroxy(((3S,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)phosphoryl)oxy)oxidophosphoryl)oxy)methyl)tetrahydrofuran-2-yl)-2-imino-3,9-dihydro-2H-purin-6-olate

Inchi Code

1S/C16H25N5O16P2.2Na/c17-16-19-12-6(13(28)20-16)18-3-21(12)14-10(26)8(24)5(34-14)2-33-38(29,30)37-39(31,32)36-15-11(27)9(25)7(23)4(1-22)35-15;;/h3-5,7-11,14-15,22-27H,1-2H2,(H,29,30)(H,31,32)(H3,17,19,20,28);;/q;2*+1/p-2/t4-,5-,7-,8-,9+,10-,11+,14-,15?;;/m1../s1

InChI key

XOAGKSFNHBWACO-NJLPRVPTSA-L

storage temp.

Sealed in dry, store in freezer, under -20C

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/103301-73-1.html

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.ambeed.com/products/103301-73-1.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library