Skip to Content
MilliporeSigma

Skip To

AMBH9884C373

Ambeed, Inc.

Timolol Related Compound B

Synonym(s):

3-(tert-butylamino)-2-(4-morpholino-1,2,5-thiadiazol-3-yloxy)propan-1-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
50 mg
Ships in 5 days - from Aldrich Partner.
$188.60
100 mg
Ships in 5 days - from Aldrich Partner.
$318.55
250 mg
Ships in 5 days - from Aldrich Partner.
$635.95
1 g
Ships in 5 days - from Aldrich Partner.
$1,906.70

About This Item

Linear Formula:
C13H24O3N4S1
CAS Number:
Molecular Weight:
316.42
MDL number:

Vendor SKU: A283279-50MG

$188.60


Ships in 5 days - from Aldrich Partner.

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C13H24N4O3S/c1-13(2,3)14-8-10(9-18)20-12-11(15-21-16-12)17-4-6-19-7-5-17/h10,14,18H,4-9H2,1-3H3

Inchi Key

MAMSPDKEMJALRZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

solid



hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library