Skip to Content
MilliporeSigma

AMBH9884C83B

Ambeed, Inc.

3-Methyl-2-(trimethylsilyl)pyridine

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Product AMBH9884C83B is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
165.31
MDL number:

Skip To

Marketplace Support
Need help or have questions about this product?
Contact Seller

Iupac Name

3-methyl-2-(trimethylsilyl)pyridine

Inchi Code

1S/C9H15NSi/c1-8-6-5-7-10-9(8)11(2,3)4/h5-7H,1-4H3

InChI key

QTQFJXMQUVUXKI-UHFFFAOYSA-N

storage temp.

Inert atmosphere, room temperature

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.ambeed.com/products/13737-07-0.html

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library