Skip to Content
MilliporeSigma

AMBH99C01A7D

Ambeed, Inc.

Ascomycin

Synonym(s):

(3S,4R,5S,8R,12S,14S,15R,16S,18R,19R,26aS,E)-8-ethyl-5,19-dihydroxy-3-((Z)-1-((1R,3R,4R)-4-hydroxy-3-methoxycyclohexyl)prop-1-en-2-yl)-14,16-dimethoxy-4,10,12,18-tetramethyl-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-3H-15,19-epoxypyrido[2,1-c][1]oxa[4]azacyclotricosine-1,7,20,21(4H,23H)-tetraone

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
Molecular Weight:
792.02
MDL number:

Inchi Code

1S/C43H69NO12/c1-10-30-18-24(2)17-25(3)19-36(53-8)39-37(54-9)21-27(5)43(51,56-39)40(48)41(49)44-16-12-11-13-31(44)42(50)55-38(28(6)33(46)23-34(30)47)26(4)20-29-14-15-32(45)35(22-29)52-7/h18,20,25,27-33,35-39,45-46,51H,10-17,19,21-23H2,1-9H3/b24-18-,26-20+/t25-,27+,28+,29-,30+,31-,32+,33-,35+,36-,37-,38+,39+,43+/m0/s1

Inchi Key

ZDQSOHOQTUFQEM-PKUCKEGBSA-N

Storage temp.

Inert atmosphere, store in freezer, under -20C

Purity

96%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

MSDS

https://www.ambeed.com/products/104987-12-4.html

hstatements

signalword

Danger

pictograms

Skull and crossbones

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library