Skip to Content
MilliporeSigma

AMBH9A626293

Ambeed, Inc.

IN-1130

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
420.47
MDL number:

Iupac Name

3-{[5-(6-methyl-2-pyridinyl)-4-(6-quinoxalinyl)-1H-imidazol-2-yl]methyl}benzamide

Inchi Code

1S/C25H20N6O/c1-15-4-2-7-20(29-15)24-23(17-8-9-19-21(14-17)28-11-10-27-19)30-22(31-24)13-16-5-3-6-18(12-16)25(26)32/h2-12,14H,13H2,1H3,(H2,26,32)(H,30,31)

InChI key

RYKSGWSKILPDDY-UHFFFAOYSA-N

storage temp.

Sealed in dry, 2-8C

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/868612-83-3.html

Synonym

Benzamide, 3-[[5-(6-methyl-2-pyridinyl)-4-(6-quinoxalinyl)-1H-imidazol-2-yl]methyl]-

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.ambeed.com/products/868612-83-3.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library