Skip to Content
MilliporeSigma

AMBH9A9AD5F0

Ambeed, Inc.

(Chloromethyl)ethenyl-Benzene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$11.50
25 G
$24.15

Vendor SKU: A565292-5G

$11.50


Ships in 7 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
152.62
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Iupac Name

1-(chloromethyl)-2-vinylbenzene

Inchi Code

1S/C9H9Cl/c1-2-8-5-3-4-6-9(8)7-10/h2-6H,1,7H2

InChI key

KKLSEIIDJBCSRK-UHFFFAOYSA-N

storage temp.

Inert atmosphere, store in freezer, under -20C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.ambeed.com/products/30030-25-2.html

Synonym

1-(chloromethyl)-2-vinylbenzene

Signal Word

Danger

Hazard Statements

MSDS

https://www.ambeed.com/products/30030-25-2.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service