Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AMBH9A9ADA8C

Ambeed, Inc.

Colistin sulfate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
1267.56
MDL number:
Product AMBH9A9ADA8C is currently not available for purchase. Contact Technical Service

Iupac Name

(R)-N-((S)-4-amino-1-(((2S,3R)-1-(((S)-4-amino-1-oxo-1-(((3S,6S,9S,12S,15R,18S,21S)-6,9,18-tris(2-aminoethyl)-3-((R)-1-hydroxyethyl)-12,15-diisobutyl-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptaazacyclotricosan-21-yl)amino)butan-2-yl)amino)-3-hydroxy-1-oxobutan-2-yl)amino)-1-oxobutan-2-yl)-6-methyloctanamide sulfate

Inchi Code

1S/C53H100N16O13.H2O4S/c1-9-30(6)12-10-11-13-41(72)60-33(14-20-54)48(77)69-43(32(8)71)53(82)65-36(17-23-57)45(74)64-38-19-25-59-52(81)42(31(7)70)68-49(78)37(18-24-58)62-44(73)34(15-21-55)63-50(79)39(26-28(2)3)67-51(80)40(27-29(4)5)66-46(75)35(16-22-56)61-47(38)76;1-5(2,3)4/h28-40,42-43,70-71H,9-27,54-58H2,1-8H3,(H,59,81)(H,60,72)(H,61,76)(H,62,73)(H,63,79)(H,64,74)(H,65,82)(H,66,75)(H,67,80)(H,68,78)(H,69,77);(H2,1,2,3,4)/t30-,31-,32-,33+,34+,35+,36+,37+,38+,39+,40-,42+,43+;/m1./s1

InChI key

ZJIWRHLZXQPFAD-FPSDIOKYSA-N

storage temp.

Inert atmosphere, 2-8C

Purity

19000

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/1264-72-8.html

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.ambeed.com/products/1264-72-8.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library