Skip to Content
MilliporeSigma

AMBH9ABD466F

Ambeed, Inc.

3-Phenyl-1-propyne

Synonym(s):

2-propynylbenzene

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C9H8
CAS Number:
Molecular Weight:
116.16
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C9H8/c1-2-6-9-7-4-3-5-8-9/h1,3-5,7-8H,6H2

Inchi Key

NGKSKVYWPINGLI-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

liquid

signalword

Danger

pictograms

FlameExclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service