All Photos(1)

AOB640092365

AOBChem

3,6-Difluoro-2-methoxyphenylboronic acid

CAS Number:
Molecular Weight:
187.94
MDL number:

Iupac Name

3,6-difluoro-2-methoxyphenylboronic acid

Inchi Code

1S/C7H7BF2O3/c1-13-7-5(10)3-2-4(9)6(7)8(11)12/h2-3,11-12H,1H3

InChI key

AXJIPGDXAIMBDX-UHFFFAOYSA-N

storage temp.

2-8*C

Purity

97

Country of Origin

CN

Shipping Temperature

Room Temperature

Physical Form

Solid

Product Link

https://aobchem.com/3-6-difluoro-2-methoxyphenylboronic-acid.html

Hazard Statements

H315 H319 H335

MSDS

https://aobchem.com/pub/media/files/sds/14161.pdf

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service