MilliporeSigma
All Photos(1)

AOBH97EBA5E1

AOBChem

(2,5-Dimethylphenyl)methanol

CAS Number:
Molecular Weight:
136.19

Iupac Name

(2,5-dimethylphenyl)methanol

Inchi Code

1S/C9H12O/c1-7-3-4-8(2)9(5-7)6-10/h3-5,10H,6H2,1-2H3

InChI key

LEBQTCCCNMTXSF-UHFFFAOYSA-N

Purity

95

Country of Origin

CN

Product Link

https://aobchem.com/2-5-dimethylphenyl-methanol.html

MSDS

https://aobchem.com/pub/media/files/sds/36353.pdf

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service