Skip to Content
MilliporeSigma

APO455826575

Apollo Scientific Ltd

1H-1,2,4-Triazole-3-carboxamide

Synonym(s):

1H-1,2,4-triazole-3-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Product APO455826575 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
112.09
MDL number:

Skip To

bp

315

Inchi Code

1S/C3H4N4O/c4-2(8)3-5-1-6-7-3/h1H,(H2,4,8)(H,5,6,7)

Inchi Key

ZEWJFUNFEABPGL-UHFFFAOYSA-N

Storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid-Powder

hstatements

signalword

Warning

pictograms

Exclamation mark

MSDS

https://store.apolloscientific.co.uk/product/1h-124-triazole-3-carboxamide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library