MilliporeSigma
All Photos(1)

APO455837429

Apollo Scientific Ltd

2,5-Bis(trifluoromethyl)benzoyl chloride

CAS Number:
Molecular Weight:
276.57
MDL number:

mp

74

density

1.52

Iupac Name

2,5-bis(trifluoromethyl)benzoyl chloride

Inchi Code

1S/C9H3ClF6O/c10-7(17)5-3-4(8(11,12)13)1-2-6(5)9(14,15)16/h1-3H

InChI key

LRJNPOCQOZWIGR-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/25-bistrifluoromethylbenzoyl-chloride

Flash Point

185

Melting Point

74

density

1.52

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/25-bistrifluoromethylbenzoyl-chloride

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service