Skip to Content
MilliporeSigma

APO456788133

Apollo Scientific Ltd

1-(4-Nitrophenyl)glycerol

Synonym(s):

1-(4-nitrophenyl)-1,2,3-propanetriol, 1-(4-Nitrophenyl)-1,2,3-propanetriol, PNPG

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C9H11O5N1
CAS Number:
Molecular Weight:
213.19
MDL number:

Inchi Code

1S/C9H11NO5/c11-5-8(12)9(13)6-1-3-7(4-2-6)10(14)15/h1-4,8-9,11-13H,5H2

Inchi Key

IUZVZBIQZKBWCC-UHFFFAOYSA-N

Country of Origin

GB


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library