Skip to Content
MilliporeSigma

APO456788217

Apollo Scientific Ltd

4-Nitrophenyl-alpha-L-fucopyranoside

Synonym(s):

4-nitrophenyl 6-deoxy-alpha-L-galactopyranoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
285.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H15NO7/c1-6-9(14)10(15)11(16)12(19-6)20-8-4-2-7(3-5-8)13(17)18/h2-6,9-12,14-16H,1H3/t6-,9+,10+,11-,12-/m0/s1

Inchi Key

YILIDCGSXCGACV-SQKFTNEHSA-N

Country of Origin

GB



MSDS

https://store.apolloscientific.co.uk/product/4-nitrophenyl-alpha-l-fucopyranoside



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library