Skip to Content
MilliporeSigma

APO456788256

Apollo Scientific Ltd

6-Chloro-3-indolyl-beta-D-galactopyranoside

Synonym(s):

6-chloro-1H-indol-3-yl beta-D-galactopyranoside, Salmon-Gal

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
Molecular Weight:
329.74
MDL number:

Inchi Code

1S/C14H16ClNO6/c15-6-1-2-7-8(3-6)16-4-9(7)21-14-13(20)12(19)11(18)10(5-17)22-14/h1-4,10-14,16-20H,5H2/t10-,11+,12+,13-,14-/m1/s1

Inchi Key

OQWBAXBVBGNSPW-MBJXGIAVSA-N

Purity

98%

Country of Origin

GB

MSDS

https://store.apolloscientific.co.uk/product/6-chloro-3-indolyl-beta-d-galactopyranoside


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library