Skip to Content
MilliporeSigma

APO456788792

Apollo Scientific Ltd

Streptavidin

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Product APO456788792 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
378.72
MDL number:

Skip To

Marketplace Support
Need help or have questions about this product?
Contact Seller

Iupac Name

1-(((carboxymethyl)thio)(3-chlorophenyl)methylene)piperidin-1-ium bromide

Inchi Code

1S/C14H16ClNO2S.BrH/c15-12-6-4-5-11(9-12)14(19-10-13(17)18)16-7-2-1-3-8-16;/h4-6,9H,1-3,7-8,10H2;1H

InChI key

RTWACOLFHOBGCE-UHFFFAOYSA-N

Country of Origin

GB

Product Link

https://store.apolloscientific.co.uk/product/streptavidin

MSDS

https://store.apolloscientific.co.uk/product/streptavidin


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library