Skip to Content
MilliporeSigma

APO497443943

Apollo Scientific Ltd

Ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulphonate

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Product APO497443943 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
235.2
MDL number:

Skip To

Marketplace Support
Need help or have questions about this product?
Contact Seller

bp

285-295

Iupac Name

ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate

Inchi Code

1S/C6H3FN2O4S.H3N/c7-3-1-2-4(14(10,11)12)6-5(3)8-13-9-6;/h1-2H,(H,10,11,12);1H3

Inchi Key

JXLHNMVSKXFWAO-UHFFFAOYSA-N

storage temp.

Store at -20C

Country of Origin

GB

Shipping Temperature

Shipped with Ice Packs

Physical Form

Solid-Powder

Product Link

https://store.apolloscientific.co.uk/product/ammonium-7-fluoro-213-benzoxadiazole-4-sulphonate

Synonym

Ammonium 7-fluorobenzofurazan-4-sulphonate, 7-Fluorobenzofurazan-4-sulphonic acid, ammonium salt

Boiling point

285-295

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/ammonium-7-fluoro-213-benzoxadiazole-4-sulphonate


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library