MilliporeSigma
All Photos(1)

APOH0387296B

Apollo Scientific Ltd

2,2',3,3',5,5',6,6'-Octafluoro-4,4'-biphenol monohydrate

CAS Number:
Molecular Weight:
348.15
MDL number:

bp

209-211

Iupac Name

2,2',3,3',5,5',6,6'-octafluoro[1,1'-biphenyl]-4,4'-diol hydrate

Inchi Code

1S/C12H2F8O2.H2O/c13-3-1(4(14)8(18)11(21)7(3)17)2-5(15)9(19)12(22)10(20)6(2)16;/h21-22H;1H2

InChI key

TYCYMOVGXPVZGN-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Product Link

https://store.apolloscientific.co.uk/product/22335566-octafluoro-44-biphenol-monohydrate

Boiling point

209-211

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

H315;H319;H335

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/22335566-octafluoro-44-biphenol-monohydrate

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service