Skip to Content
MilliporeSigma

APOH11A9D0A0

Apollo Scientific Ltd

Ethyl 4-hydroxy-3-methoxycinnamate

Synonym(s):

ethyl (2E)-3-(4-hydroxy-3-methoxyphenyl)-2-propenoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
222.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


bp

63-65

Inchi Code

1S/C12H14O4/c1-3-16-12(14)7-5-9-4-6-10(13)11(8-9)15-2/h4-8,13H,3H2,1-2H3

Inchi Key

ATJVZXXHKSYELS-UHFFFAOYSA-N

Storage temp.

Ambient Temperature

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Flash Point

164-166/0.5mm



hstatements

signalword

Warning

pictograms

Exclamation mark

MSDS

https://store.apolloscientific.co.uk/product/ethyl-4-hydroxy-3-methoxycinnamate



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library