Skip to Content
MilliporeSigma

APOH11A9DBB1

Apollo Scientific Ltd

1H-Pyrazole-1-carboxamidine, N,N'-Bis-BOC protected

Synonym(s):

(E)-tert-butyl (1H-pyrazol-1-yl)methanediylidenedicarbamate

Sign In to View Organizational & Contract Pricing.

Select a Size

Product APOH11A9DBB1 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
310.35
MDL number:

Skip To

bp

86-90

Inchi Code

1S/C14H22N4O4/c1-13(2,3)21-11(19)16-10(18-9-7-8-15-18)17-12(20)22-14(4,5)6/h7-9H,1-6H3,(H,16,17,19,20)

Inchi Key

QFNFDHNZVTWZED-UHFFFAOYSA-N

Storage temp.

Ambient Temperature

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

hstatements

signalword

Warning

pictograms

Exclamation mark

MSDS

https://store.apolloscientific.co.uk/product/1h-pyrazole-1-carboxamidine-nn-bis-boc-protected


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library