Skip to Content
MilliporeSigma

APOH14DBAC0E

Apollo Scientific Ltd

(+)-Camphorsulfonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

Product APOH14DBAC0E is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
232.3
MDL number:

Skip To

bp

196-200

Iupac Name

((1R,4R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonic acid

Inchi Code

1S/C10H16O4S/c1-9(2)7-3-4-10(9,8(11)5-7)6-15(12,13)14/h7H,3-6H2,1-2H3,(H,12,13,14)/t7-,10+/m1/s1

InChI key

MIOPJNTWMNEORI-XCBNKYQSSA-N

storage temp.

Ambient Temperature

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/camphorsulfonic-acid

Boiling point

196-200

pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/camphorsulfonic-acid


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library