Skip to Content
MilliporeSigma

ATE014416409

AstaTech, Inc.

3-BROMO PHENOL

Synonym(s):

3-bromophenol

Sign In to View Organizational & Contract Pricing.

Select a Size

25 G

$23.75

100 G

$61.25

500 G

$223.75

Vendor SKU: 26922-25G

$23.75


Ships in 10 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
173.01
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

bp

235-236 C

mp

28-32 C

Inchi Code

1S/C6H5BrO/c7-5-2-1-3-6(8)4-5/h1-4,8H

Inchi Key

MNOJRWOWILAHAV-UHFFFAOYSA-N

Storage temp.

2-8 C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Pale-yellow to Yellow-brown Solid

Flash Point

>110 C/230 F

hstatements

signalword

Warning


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service