All Photos(1)

ATE386054359

AstaTech, Inc.

8-BROMOISOQUINOLIN-3-AMINE

CAS Number:
Molecular Weight:
223.07
MDL number:

Iupac Name

8-bromo-3-isoquinolinylamine

Inchi Code

1S/C9H7BrN2/c10-8-3-1-2-6-4-9(11)12-5-7(6)8/h1-5H,(H2,11,12)

InChI key

MELBFTIXTXTDDR-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=14435

Signal Word

Warning

Hazard Statements

H302;H315;H320;H335

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=68517

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service