Skip to Content
MilliporeSigma

ATE389310587

AstaTech, Inc.

2'-FLUORO-N4-ACETYL-5'-O-DMT-2'-DEOXYCYTIDINE-3'-CE-PHOSPHORAMIDITE

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$115.00
5 G
$356.25

Vendor SKU: HG1235-1G

$115.00


Ships in 10 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
705.68
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Iupac Name

(2R,3R,4R,5R)-5-(4-acetamido-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-fluorotetrahydrofuran-3-yl (2-cyanoethyl) phosphoramidite

Inchi Code

1S/C35H37FN5O8P/c1-23(42)39-30-18-20-41(34(43)40-30)33-31(36)32(49-50(38)47-21-7-19-37)29(48-33)22-46-35(24-8-5-4-6-9-24,25-10-14-27(44-2)15-11-25)26-12-16-28(45-3)17-13-26/h4-6,8-18,20,29,31-33H,7,21-22,38H2,1-3H3,(H,39,40,42,43)/t29-,31-,32-,33-,50?/m1/s1

InChI key

CCWQNSIALJQUHV-MPONISAVSA-N

storage temp.

?20C

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

Product Link

https://astatechinc.com/CPSResult.php?CRNO=17856

pictograms

Signal Word

Danger

Hazard Statements


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service