All Photos(1)

ATE398811338

AstaTech, Inc.

1-OXA-4,9-DIAZASPIRO[5.5]UNDECANE 2HCL

CAS Number:
Molecular Weight:
229.15
MDL number:

Iupac Name

1-oxa-4,9-diazaspiro[5.5]undecane dihydrochloride

Inchi Code

1S/C8H16N2O.2ClH/c1-3-9-4-2-8(1)7-10-5-6-11-8;;/h9-10H,1-7H2;2*1H

InChI key

DRCFFILZGMKHTP-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=17992

Signal Word

Warning

Hazard Statements

H302;H315;H319;H335

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=81382

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service