Skip to Content
MilliporeSigma

ATE428005821

AstaTech, Inc.

3,3'-DIAMINOBENZIDINE TETRAHCL DIHYDRATE

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
396.14
MDL number:
Marketplace Support
Need help or have questions about this product?
Contact Seller
Marketplace Support
Need help or have questions about this product?
Contact Seller

Iupac Name

[1,1'-biphenyl]-3,3',4,4'-tetramine tetrahydrochloride dihydrate

Inchi Code

1S/C12H14N4.4ClH.2H2O/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8;;;;;;/h1-6H,13-16H2;4*1H;2*1H2

InChI key

BXIYIGBKSGVJOS-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Solid

Product Link

https://astatechinc.com/CPSResult.php?CRNO=40737

Signal Word

Warning

Hazard Statements

Precautionary statements


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library