Skip to Content
MilliporeSigma

ATE428009591

AstaTech, Inc.

ETOPOSIDE

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Product ATE428009591 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
588.57
MDL number:

Skip To

Marketplace Support
Need help or have questions about this product?
Contact Seller

mp

236-251 C(lit.)

Iupac Name

(5S,5aS)-9-(((2S,6S,7S,8S,8aR)-7,8-dihydroxy-2-methylhexahydropyrano[3,2-d][1,3]dioxin-6-yl)oxy)-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,8,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(5aH)-one

Inchi Code

1S/C29H32O13/c1-11-36-9-20-27(40-11)24(31)25(32)29(41-20)42-26-14-7-17-16(38-10-39-17)6-13(14)21(22-15(26)8-37-28(22)33)12-4-18(34-2)23(30)19(5-12)35-3/h4-7,11,15,20-22,24-27,29-32H,8-10H2,1-3H3/t11-,15?,20?,21-,22+,24-,25-,26?,27-,29+/m0/s1

InChI key

VJJPUSNTGOMMGY-ZURUWIRVSA-N

storage temp.

2-8 C

Purity

99%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=44181

Melting Point

236-251 C(lit.)

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/CPSResult.php?CRNO=44181


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library