Skip to Content
MilliporeSigma

ATE448434813

AstaTech, Inc.

6-BROMO-2H-BENZO[B][1,4]THIAZIN-3(4H)-ONE

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
244.11
MDL number:

Iupac Name

6-bromo-2H-1,4-benzothiazin-3(4H)-one

Inchi Code

1S/C8H6BrNOS/c9-5-1-2-7-6(3-5)10-8(11)4-12-7/h1-3H,4H2,(H,10,11)

InChI key

QXMFOFHASVVZIT-UHFFFAOYSA-N

Purity

97%

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

https://astatechinc.com/CPSResult.php?CRNO=61428

Hazard Statements

Precautionary statements


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library