Skip to Content
MilliporeSigma

ATE456785799

AstaTech, Inc.

4,4'-BIPHENYLDISULFONYL CHLORIDE

Synonym(s):

[1,1'-biphenyl]-4,4'-disulfonyl dichloride

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C12H8O4Cl2S2
CAS Number:
Molecular Weight:
351.23
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C12H8Cl2O4S2/c13-19(15,16)11-5-1-9(2-6-11)10-3-7-12(8-4-10)20(14,17)18/h1-8H

Inchi Key

OTFAWEIFBPUXOH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

2-8 C

Country of Origin

CN

Physical Form

Solid

Melting Point

205-208 C(lit.)

Looking for similar products? Visit Product Comparison Guide

hstatements

signalword

Danger

pictograms


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service