Skip to Content
MilliporeSigma

ATE517248637

AstaTech, Inc.

OLIGOMYCIN A

Synonym(s):

(1S,2'S,4E,5'R,6R,6'R,7S,8R,10S,11S,12R,14S,15R,16S,18E,22S,25R,28R,29S )-22-ethyl-7,11,14,15-tetrahydroxy-6'-((S)-

Sign In to View Organizational & Contract Pricing.

Select a Size

Product ATE517248637 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
791.08
MDL number:

Skip To

Inchi Code

1S/C45H74O11/c1-12-34-17-15-13-14-16-27(4)42(51)44(11,53)43(52)32(9)40(50)31(8)39(49)30(7)38(48)26(3)18-21-37(47)54-41-29(6)35(20-19-34)55-45(33(41)10)23-22-25(2)36(56-45)24-28(5)46/h13-15,17-18,21,25-36,38,40-42,46,48,50-51,53H,12,16,19-20,22-24H2,1-11H3/b14-13+,17-15+,21-18+/t25-,26-,27+,28+,29+,30-,31-,32-,33-,34-,35-,36-,38+,40+,41+,42-,44+,45-/m1/s1

Inchi Key

MNULEGDCPYONBU-WMBHJXFZSA-N

Storage temp.

?20C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

hstatements

signalword

Warning


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library