Sign In to View Organizational & Contract Pricing.
Select a Size
Product ATE517251755 is currently not available for purchase. Contact Technical Service
About This Item
Skip To
bp
130-140 C/0.01 mmHg(lit.)
Inchi Code
1S/C9H14Br2O4/c1-14-8(12)6(10)4-3-5-7(11)9(13)15-2/h6-7H,3-5H2,1-2H3
Inchi Key
AWWJYEJSCIDADZ-UHFFFAOYSA-N
Purity
95%
Country of Origin
CN
Physical Form
Liquid
Flash Point
113 C/235.4 F
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.