Skip to Content
MilliporeSigma

ATE517252551

AstaTech, Inc.

1-BROMOADAMANTANE

Synonym(s):

(3s,5s,7s)-1-bromoadamantane

Sign In to View Organizational & Contract Pricing.

Select a Size

25 G

$27.50

Vendor SKU: O10392-25G

$27.50


Ships in 10 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
215.13
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

mp

116-118 C(lit.)

Inchi Code

1S/C10H15Br/c11-10-4-7-1-8(5-10)3-9(2-7)6-10/h7-9H,1-6H2/t7-,8+,9-,10?

Inchi Key

VQHPRVYDKRESCL-URIMCQPQSA-N

Storage temp.

Room temperature

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Pale-yellow to Yellow-brown Solid

hstatements

signalword

Warning


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service