MilliporeSigma
All Photos(1)

ATE638636221

AstaTech, Inc.

2-(1-METHYL-1H-IMIDAZOL-4-YL)ACETIC ACID HCL

CAS Number:
Molecular Weight:
176.6
MDL number:

Iupac Name

(1-methyl-1H-imidazol-4-yl)acetic acid hydrochloride

Inchi Code

1S/C6H8N2O2.ClH/c1-8-3-5(7-4-8)2-6(9)10;/h3-4H,2H2,1H3,(H,9,10);1H

InChI key

XVLSBASXRIOQRZ-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Yellow to Brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=101927

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=P16507

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service